Skip to Content
Merck

1.07237

BPLS agar

for the isolation of Salmonella

Synonym(s):

Brilliant-green phenol-red lactose sucrose agar, Brillant-green phenol-red lactose sucrose agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106213
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

medium granules (dehydrated (DCM))

technique(s)

microbiological culture: suitable

pH

6.7-7.1 (25 °C in H2O)

solubility

52 g/L

bulk density

700 kg/m3

application(s)

clinical
microbiology

suitability

Salmonella spp.

General description

for the isolation of Salmonella

Analysis Note

Appearance (clearness)clearAppearance (color)redSolidification behaviour (2 hrs., 45 °C)liquidpH-value (25 °C)6.7 - 7.1Recovery on test medium (Salmonella typhimurium ATCC 14028 (WDCM 00031))≥ 70 %Recovery on test medium (Salmonella choleraesuis ATCC 13312)≥ 70 %Recovery on test medium (Salmonella enteritidis NCTC 5188 (WDCM 00119))≥ 70 %Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013))Recovery on test medium (Proteus hauseri ATCC 13315)Recovery on test medium (Staphylococcus aureus ATCC 25923 (WDCM 00034))Recovery on test medium (Enterococcus faecalis ATCC 33186)Recovery on test medium (Bacillus subtilis ATCC 6633 (WDCM 00003))Colony color (Salmonella typhimurium ATCC 14028 (WDCM 00031))pinkColony color (Salmonella choleraesuis ATCC 13312)pinkColony color (Salmonella enteritidis NCTC 5188 (WDCM 00119))pinkColony color (Escherichia coli ATCC 25922 (WDCM 00013))yellowColony color (Proteus hauseri ATCC 13315)yellowColony color (Staphylococcus aureus ATCC 25923 (WDCM 00034))yellowColony color (Enterococcus faecalis ATCC 33186)yellowColony color (Bacillus subtilis ATCC 6633 (WDCM 00003))yellow/orangeIncubation: 24 hrs.; 35 °C; aerobically.


Still not finding the right product?

Explore all of our products under BPLS agar


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library