Skip to Content
Merck
All Photos(2)

Key Documents

1.07179

Supelco

n-Pentane

for spectroscopy Uvasol®

Synonym(s):

Pentane

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€298.00

€298.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€298.00

About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-692-4
NACRES:
NA.03
technique(s):
UV/Vis spectroscopy: suitable
bp:
35-36 °C (lit.)

€298.00


Estimated to ship on30 May 2025


Request a Bulk Order

vapor density

2.48 (vs air)

Quality Level

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

500 °F

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

~8.3 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.005% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

200 nm, ≥50%
210 nm, ≥70%
215 nm, ≥85%
225 nm, ≥95%
240 nm, ≥98%

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

transition temp

flash point -48 °C (refers to pure substance)

solubility

0.4 g/L

density

0.626 g/mL at 25 °C (lit.)

UV absorption

λ: 200 nm Amax: ≤0.30
λ: 210 nm Amax: ≤0.16
λ: 215 nm Amax: ≤0.07
λ: 225 nm Amax: ≤0.02
λ: 240 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application


  • Quantitative determination of nitrous oxide in human blood by HS-GC-MS: forensic application of two fatal poisoning cases.: This study leverages n-Pentane in headspace gas chromatography-mass spectrometry (HS-GC-MS) for the forensic analysis of nitrous oxide in human blood. Highlighting its importance in forensic science, the research demonstrates n-Pentane′s utility in providing accurate and reliable measurements essential for legal and medical evaluations (Li Z et al., 2024).

  • A photochromic trinuclear dysprosium(iii) single-molecule magnet with two distinct relaxation processes.: This article describes the use of n-Pentane in the synthesis of a dysprosium-based single-molecule magnet, showcasing its role in developing materials with unique magnetic properties for potential applications in quantum computing and data storage (Rogacz K et al., 2024).

  • Reduction of Thorium Tris(amido)arene Complexes: Reversible Double and Single C-C Couplings.: This research employs n-Pentane in the study of thorium complexes, providing insights into reversible carbon-carbon coupling reactions. The findings contribute to the understanding of actinide chemistry and its application in recycling and waste reduction strategies (Deng C et al., 2024).

  • Tuning Reactivities of tert-Butyllithium by the Addition of Stoichiometric Amounts of Tetrahydrofuran.: The study utilizes n-Pentane to adjust the reactivity of tert-butyllithium, illustrating its significant role in enhancing the control over chemical reactions. This advancement is pivotal for the synthesis of complex molecules in pharmaceuticals and materials science (Kleinheider J et al., 2024).

  • Steps to achieve carvone-rich spearmint (Mentha spicata L.) essential oil: a case study on the use of different distillation methods.: In this research, n-Pentane is examined for its efficacy in extracting high-quality essential oils from spearmint, demonstrating its versatility in botanical extractions and its impact on the quality and characteristics of the essential oils obtained (Moradi-Sadr J et al., 2023).

Analysis Note

Purity (GC): ≥ 99.5 %
Evaporation residue: ≤ 0.0002 %
Water: ≤ 0.005 %
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 1.0 ppb
Transmission (at 200 nm): ≥ 50 %
Transmission (at 210 nm): ≥ 70 %
Transmission (at 215 nm): ≥ 85 %
Transmission (at 225 nm): ≥ 95 %
Transmission (from 240 nm): ≥ 98 %
Absorbance (at 200 nm): ≤ 0.30
Absorbance (at 210 nm): ≤ 0.16
Absorbance (at 215 nm): ≤ 0.07
Absorbance (at 225 nm): ≤ 0.02
Absorbance (from 240 nm): ≤ 0.01
Meet the requirements of "Specific use spectroscopy" according Reag. Ph Eur

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service