Skip to Content
Merck
All Photos(1)

Key Documents

1.07160

Supelco

Paraffin

viscous Reag. Ph Eur

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€69.90
2.5 L
€148.00
25 L
€362.00
180 L
€2,090.00

€69.90


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 L
€69.90
2.5 L
€148.00
25 L
€362.00
180 L
€2,090.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181503
EC Index Number:
232-455-8
NACRES:
NB.51

€69.90


Estimated to ship on01 June 2025


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

form

liquid

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

technique(s)

cell culture | mammalian: suitable

dynamic viscosity

100-230 mPa.s(20 °C)

kinematic viscosity

>22.5 cSt(40 °C)

bp

300-500 °C/1,013 hPa

transition temp

flash point 200 °C

density

0.86 g/cm3 at 20 °C

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Identity (IR-spectrum): passes test
Identity (Viscosity): passes test
Acidity or alkalinity: passes test
Relative Density (d 20/20): 0.827 - 0.890
Dynamic viscosity (20 °C): 110 - 230 mPa∙s
Aromatic, polycyclic Hydrocarbons: passes test
Solid Paraffines: passes test
Readily carbonisable substance: passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

455.0 °F

Flash Point(C)

235 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service