Skip to Content
Merck
All Photos(1)

Key Documents

1.07150

Supelco

Paraffin

42-44, in block form

Synonym(s):

Paraffin

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€139.00
2.5 KG
€294.00
25 KG
€851.00

€139.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€139.00
2.5 KG
€294.00
25 KG
€851.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181503
EC Index Number:
232-315-6
NACRES:
NB.51

€139.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

form

solid

autoignition temp.

>300 °C

potency

>5000 mg/kg LD50, oral (Rat)
>3600 mg/kg LD50, skin (Rabbit)

impurities

≤0.015% Sulfate (SO4)

pH

7

bp

>300 °C/1013 hPa

mp

42-72 °C

density

0.90 g/cm3 at 20 °C

storage temp.

2-30°C

General description

42-44, in block form

Analysis Note

Identity (IR-spectrum)passes testAcidity or alkalinitypasses testSolidification point42 - 44 °CSulfate (SO₄)≤ 0.015 %

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service