Skip to Content
Merck
All Photos(2)

Key Documents

1.07043

Millipore

Mixed Peptone for Biotechnology and Microbiology

Prime, for Biotechnology and Microbiology

Synonym(s):

Poly peptone, peptone mixture

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€607.00

€607.00


Estimated to ship on26 May 2025


Request more information

Select a Size

Change View
1 KG
€607.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

€607.00


Estimated to ship on26 May 2025


Request more information

Product Name

Universal peptone M 66, suitable for microbiology

form

powder

Quality Level

packaging

pkg of 1 kg

pH

6-7 (20 g/L in H2O)

solubility

490 g/L

bulk density

370 kg/m3

application(s)

microbiology

storage temp.

15-25°C

General description

Universal M 66 peptone is a poly peptone mixture of casein peptone, meat peptone and soy peptone, with well-balanced nutritional characteristics. Peptone M 66 is widely used in industrial microbiology as a nutrient source for the growth of microorganisms in large-scale fermentation processes. Peptone M 66 is a water-soluble protein hydrolysate obtained by the digestion of natural sources such as meat, casein, gelatin, or soy. It provides a variety of essential nutrients required for bacterial growth, including amino acids, vitamins, and minerals. It is useful for the growth of fastidious bacteria, yeasts, and molds.

Application

Universal peptone M 66 can be used as nutrient, for example as replacement of serum albumin in cell culture and fermentation. It is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts, and molds from various pharmaceutical, environmental, and food and beverage samples.

Analysis Note

Peptone content (ex N, calc. on dried substance): ≥ 80 %
Amino nitrogen (as N): ≥ 3.5 %
pH-value (2 %; water): 6 - 7
Sulfated ash (800 °C): ≤ 15 %
Heavy metals (as Pb): ≤ 0.001 %
Loss on Drying (105°C): ≤ 5 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service