Skip to Content
Merck
All Photos(1)

Documents

1.06595

Supelco

Rhodizonic acid disodium salt

indicator for sulfate titration

Synonym(s):

Rhodizonic acid disodium salt, 1,2-Dihydroxy-3,4,5,6-tetraoxo-1-cyclohexene disodium salt, Sodium rhodizonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Na2O6
CAS Number:
Molecular Weight:
214.04
MDL number:
UNSPSC Code:
12352107
EC Index Number:
208-340-3

Quality Level

form

solid

mp

>300 °C

bulk density

620 kg/m3

storage temp.

15-25°C

InChI

1S/C6O6.2Na/c7-1-2(8)4(10)6(12)5(11)3(1)9;;/q-2;2*+1

InChI key

WNLGVLVRNGQRFD-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 98.5 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax.(Hydrochloric acid 0.1 mol/l): 317 - 320 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.02 g/l; hydrochloric acid 0.1 mol/l): ≥ 540
Suitability as indicator (for sulfate titration): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service