Skip to Content
Merck
All Photos(1)

Key Documents

1.06572

Supelco

tri-Sodium phosphate dodecahydrate

for analysis EMSURE®

Synonym(s):

tri-Sodium phosphate dodecahydrate, Sodium phosphate tribasic, Trisodium phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€60.60
5 KG
€212.00
25 KG
€1,210.00

€60.60


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€60.60
5 KG
€212.00
25 KG
€1,210.00

About This Item

Empirical Formula (Hill Notation):
Na3O4P · 12H2O
CAS Number:
Molecular Weight:
380.12
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-509-8
NACRES:
NA.21
form:
crystals
solubility:
285 g/L

€60.60


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

product line

EMSURE®

Assay

98.0-102.0% (alkalimetric)

form

crystals

potency

7400 mg/kg LD50, oral (Rat)

impurities

≤0.001% Total nitrogen (N)
≤0.01% Insoluble matter
≤2.5% Free alkali (as NaOH)

pH

12 (20 °C, 10 g/L in H2O)

mp

75 °C (Elimination of water of crystallisation)

solubility

285 g/L

density

1.62 g/cm3 at 20 °C

bulk density

950 kg/m3

anion traces

chloride (Cl-): ≤0.005%
fluoride (F-): ≤0.0005%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
excess alkali (as NaOH): ≤2.5%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[Na+].[P](=O)([O-])([O-])[O-].O.O.O.O.O.O.O.O.O.O.O.O

InChI

1S/3Na.H3O4P.12H2O/c;;;1-5(2,3)4;;;;;;;;;;;;/h;;;(H3,1,2,3,4);12*1H2/q3*+1;;;;;;;;;;;;;/p-3

InChI key

ASTWEMOBIXQPPV-UHFFFAOYSA-K

Analysis Note

Assay (alkalimetric): 98.0 - 102.0
Identity: passes test
Insoluble matter: ≤ 0.01 %
Free alkali (as NaOH): ≤ 2.5 %
Chloride (Cl): ≤ 0.005 %
Fluoride (F): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service