Skip to Content
Merck
All Photos(1)

Key Documents

1.06062

Sigma-Aldrich

N,N′-Methylenebisacrylamide ready-to-use solution "BIS" 2%

for electrophoresis

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€105.00

€105.00


Estimated to ship on25 April 2025



Select a Size

Change View
1 L
€105.00

About This Item

UNSPSC Code:
41105323
NACRES:
NC.01

€105.00


Estimated to ship on25 April 2025


Quality Level

form

liquid

density

1.00 g/cm3 at 20 °C

storage temp.

2-8°C

Analysis Note

Assay (by acidimetry following saponification): 1.80 - 2.20 %
Absorbance A (290 nm; 2.0 %; 1 cm): ≤ 0.4

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service