Skip to Content
Merck

1.06008

SAFC

Methanol

EMPROVE® ESSENTIAL, Ph. Eur., BP, JPE, NF

Pharma Manufacturing

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€33.00
2.5 L
€47.00
10 L
€332.00
25 L
€439.00
180 L
Inquire
190 L
Inquire
950 L
Inquire

€33.00


Estimated to ship on02 June 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 L
€33.00
2.5 L
€47.00
10 L
€332.00
25 L
€439.00
180 L
Inquire
190 L
Inquire
950 L
Inquire

About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-659-6

€33.00


Estimated to ship on02 June 2025


Request a Bulk OrderRequest a Sample

Agency

BP
JPE
NF
Ph. Eur.

Quality Level

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

API processing | polymorph screening: suitable

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Methanol is used as solvent in a variety of pharmaceutical production process, including tablet coating or microsphere production.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service