Skip to Content
Merck
All Photos(1)

Key Documents

1.05708

Supelco

Molecular sieve 0.4 nm

beads ~ 2 mm / ~ 10 mesh Reag. Ph Eur

Synonym(s):

Molecular sieve 0.4 nm, Sodium aluminium silicate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€125.00
1 KG
€271.00
10 KG
€2,180.00

€125.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
250 G
€125.00
1 KG
€271.00
10 KG
€2,180.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41123003
EC Index Number:
215-283-8
NACRES:
NB.21

€125.00


Estimated to ship on02 June 2025


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

form

beads

particle size

~10 mesh
~2 mm

pore size

0.4 nm

pH

8-11 ( in H2O)

mp

>1600 °C

bulk density

700‑750 kg/m3

storage temp.

no temp limit

SMILES string

[Si](=O)=O.[Al](=O)O[Al]=O

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

General description

Molecular sieve 0.4 nm is a porous solid that can be used in desiccators and drying tubes to dry gases and absolute solvents. It also changes its color from brown to yellowish at a water uptake of approximately 7-10 g/100 g molecular sieve.

Application

Molecular sieve 0.4 nm can be used as a catalyst:
  • For the conversion of epoxides to thiiranes by treating with thiourea.
  • In the solvent-free acetylation of eugenol with acetic anhydride to synthesize eugenyl acetate.
It can also be used as catalyst support in the copper-palladium bimetallic catalyzed Suzuki-Miyaura coupling of aryl halides with phenylboronic acids to synthesize biphenyls.

Features and Benefits

  • Easy to use
  • Chemically inert
  • Non-toxic
  • Dried liquids can be decanted
  • High adsorption capacity
  • Selective adsorption
  • Reusable

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service