Skip to Content
Merck
All Photos(1)

Key Documents

1.05271

Millipore

Antibiotic agar No. 5

suitable for microbiology

Synonym(s):

Antibiotic agar No 5 (GROVE and RANDALL), GROVE and RANDALL medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

powder (dehydrated (DCM))

Quality Level

manufacturer/tradename

GranuCult®

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

Micrococcus spp.
bacteria

General description

for microbiology

Analysis Note

Appearance (clearness): clear to slightly turbid
Appearance (colour): yellowish-brown
pH-value (25 °C): 7.7 - 8.1
Solidification behaviour (2 hrs., 45 °C): liquid
Growth (Bacillus subtilis BGA): good to very good
Inhibition zone diameter/Streptomycin 0.5 µg (Bacillus subtilis BGA):
Inhibition zone diameter/Streptomycin 10 µg (Bacillus subtilis BGA):
Inhibition zone diameter/Gentamicin 10 µg (Bacillus subtilis BGA):
Inhibition zone diameter/Gentamycin 30 µg (Bacillus subtilis BGA):
Incubation: 24 hrs.; 35 °C; aerobic

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service