Skip to Content
Merck
All Photos(1)

Key Documents

1.05174

Sigma-Aldrich

Hematoxylin solution modified acc. to Gill III

for microscopy

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€76.10
1 L
€130.00
2.5 L
€233.00

€76.10


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€76.10
1 L
€130.00
2.5 L
€233.00

About This Item

UNSPSC Code:
41116124
NACRES:
NA.41

€76.10


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

pH

2.5 (20 °C in H2O)

density

1.07 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Hematoxylin solution modified according to Gill III - for microscopy, is used for human-medical cell diagnosis and serves the histological and clinico-cytological investigation of sample material of human origin. It is a ready-to-use staining solution that when used together with other in vitro diagnostic products from our portfolio makes target structures evaluable for diagnostic purposes in histological and clinico-cytological specimen materials, for example histological sections of e. g. the kidney, muscle tissue, heart, or lung. In hematoxylin-eosin staining (H&E), the most widely used histological staining technique, nuclear staining is achieved using ready-to-use staining solutions, such as Mayer′s hemalum solution or Hematoxylin solution modified according to Gill III. These solutions do not need to be filtrated prior use and the dye is oxidized in a controlled manner, which renders the results robust and allows a brilliant differentiation of the stained structures. Counterstaining of i.e. for proteins, collagen, keratin, intercellular substances. can be performed with alcoholic or aqueous Eosin Y solution 0.5 %.
A 500 ml bottle is sufficient for 1500 - 2500 stainings. It is registered as IVD and CE product. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy: passes test
Nuclei: dark blue to dark violet
Cytoplasm: pink to red
intercellular substances: pink to red
Erythrocytes: yellow to orange

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Questions

1–2 of 2 Questions  
  1. I would like to know ,what is the UN number of hematoxylin gills?

    1 answer
    1. The UN number for any product can be found Section 14 of the SDS. Product 105174 does not have an assigned UN number. The product is considered Not Dangerous Goods since the product is not considered hazardous within the framework international trade and transport.

      Helpful?

  2. Can the product be used for immunohistochemistry as a counterstain in a Biotin and DAB stain protocol without affecting the DAB stains?

    1 answer
    1. The Users Guide for product 1051740500 mentions the solution for use with a Hematoxylin and Eosin (H&E) stain procedure. Gill's 3 is a progressive hematoxylin, and slides do not typically need to be differentiated in an acidic solution to remove overstaining from the nucleus and cytoplasm. It is suggested to reduce the staining times for both Mayers Hematoxylin and Gill's formulations to achieve the desired staining.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service