Skip to Content
Merck

1.04900

SAFC

Potassium bromide

EMPROVE® EVOLVE

Synonym(s):

Potassium bromide, Hydrobromic Acid Potassium Salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€242.00
25 KG
Inquire

€242.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 KG
€242.00
25 KG
Inquire

About This Item

Empirical Formula (Hill Notation):
BrK
CAS Number:
Molecular Weight:
119.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-830-3

€242.00


Estimated to ship on21 May 2025


Request a Bulk OrderRequest a Sample

vapor pressure

1.3 hPa ( 795 °C)

Quality Level

description

EMPROVE® EVOLVE

product line

EMPROVE® EVOLVE

form

solid

pH

5.5-8.5 (20 °C, 50 g/L in H2O)

bp

1435 °C/1013 hPa

mp

730 °C

solubility

650 g/L

density

2.75 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium bromide Emprove® Essential is mainly used in highly regulated pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service