Skip to Content
Merck
All Photos(1)

Documents

1.04738

SAFC

Invertin MERCK

humectant for confectionery E 1103

Pharma Manufacturing

Synonym(s):

Humectants for confectionery

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
E Number:
E1103
EC Index Number:
232-615-7

Quality Level

form

liquid

pH

5.2-5.6 (20 °C in H2O)

density

1.16 g/cm3 at 20 °C

application(s)

food and beverages

storage temp.

2-8°C

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Invertin MERCK humectant for confectionery E 1103 is used as a food additive.

Legal Information

INVERTIN MERCK is a trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service