Skip to Content
Merck
All Photos(1)

Key Documents

1.04619

Supelco

Hydroxylammonium chloride

for analysis (≤ 0.000001% Hg) ACS,ISO

Synonym(s):

Hydroxylammonium chloride, Hydroxylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€366.00

€366.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€366.00

About This Item

Empirical Formula (Hill Notation):
H3NO · xHCl
CAS Number:
Molecular Weight:
33.03 (free base basis)
MDL number:
UNSPSC Code:
12352301
EC Index Number:
226-798-2
NACRES:
NB.21
grade:
ACS reagent
vapor pressure:
0.054 Pa ( 50 °C)

€366.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. ISO

vapor pressure

0.054 Pa ( 50 °C)

Assay

≥99% (manganometric)

form

solid

dilution

(for analytical testing)

pH

2.5-3.5 (20 °C, 50 g/L in H2O)

mp

154 °C

density

1.70 g/cm3 at 20.2 °C

bulk density

900 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].NO.[H+]

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2

InChI key

WTDHULULXKLSOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Facile synthesis of light harvesting semiconductor bismuth oxychloride nano photo-catalysts for efficient removal of hazardous organic pollutants.: This study demonstrates the use of hydroxylammonium chloride in the synthesis of bismuth oxychloride nanophotocatalysts. These catalysts are utilized for the degradation of hazardous organic compounds, highlighting a significant application in environmental remediation and analytical chemistry (Seddigi ZS et al., 2017).

  • On-column derivatization using redox activity of porous graphitic carbon stationary phase: an approach to enhancement of separation selectivity of liquid chromatography.: This article explores the use of hydroxylammonium chloride in on-column derivatization during liquid chromatographic processes. It focuses on enhancing separation selectivity, illustrating its pivotal role in refining analytical methods for complex mixtures (Shibukawa M et al., 2003).


Analysis Note

Assay (manganometric): ≥ 99.0 %
Clarity of the solution in Ethanol: passes test
pH-value (5 %; water): 2.5 - 3.5
Titrable free acid: ≤ 0.25 meq/g
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (ACS): ≤ 0.0005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Hg (Mercury): ≤ 0.000001 %
NH₄ (Ammonium): passes test
Pb (Lead): ≤ 0.0005 %
Total sulfur (as sulfate): ≤ 0.005 %
Residue on ignition (as sulfate): ≤ 0.01 %
Corresponds to ACS,ISO

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service