Skip to Content
Merck
All Photos(1)

Key Documents

1.04527

Supelco

3-Methyl-2-benzothiazolinone-hydrazonehydrochloride

GR for analysis Reag. Ph Eur

Synonym(s):

3-Methyl-2-benzothiazolinone-hydrazonehydrochloride, MBTH

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€236.00

€236.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
€236.00

About This Item

Empirical Formula (Hill Notation):
C8H10ClN3S
CAS Number:
Molecular Weight:
215.70
MDL number:
UNSPSC Code:
12352107
EC Index Number:
224-393-5
NACRES:
NA.05

€236.00


Estimated to ship on28 May 2025


Request a Bulk Order

Agency

reag. Ph. Eur.

Quality Level

form

solid

potency

149 mg/kg LD50, oral (Rat)
12300 mg/kg LD50, skin (Rabbit)

pH

3-4 (20 °C, 4 g/L in H2O)

bulk density

748 kg/m3

storage temp.

15-25°C

InChI

1S/C8H9N3S.Cl/c1-11-6-4-2-3-5-7(6)12-8(11)10-9;/h2-5H,9H2,1H3;/b10-8+;

InChI key

WAGARHVUZQWORV-VRTOBVRTSA-N

Application

  • Kinetic spectrophotometric determination of ethamsylate in dosage forms.: The research describes a kinetic spectrophotometric method for determining ethamsylate using 3-Methyl-2-benzothiazolinone-hydrazone hydrochloride. The method is efficient for analyzing ethamsylate in various pharmaceutical formulations (El-Enany et al., 2007).
  • Spectrophotometric determination of dobutamine hydrochloride using 3-methylbenzothiazolin-2-one hydrazone.: This study provides a spectrophotometric method for dobutamine hydrochloride analysis, highlighting the utility of 3-Methyl-2-benzothiazolinone-hydrazone hydrochloride in analytical chemistry for pharmaceutical applications (el-Kommos, 1987).
  • Microdetermination of D-amino acids and D-amino acid oxidase activity with 3,methyl-2-benzothiazolone hydrazone hydrochloride.: This research introduces a microdetermination method for D-amino acids and D-amino acid oxidase activity using 3-Methyl-2-benzothiazolinone-hydrazone hydrochloride, contributing to biochemical analysis techniques (Soda, 1968).

Analysis Note

Assay (argentometric): ≥ 99
Identity (IR-spectrum): passes test
pH-value (0.4 %, water): 3 - 4
Melting point: about 270
Sulfated ash: ≤ 0.1 %
Loss on Drying (105°C): ≤ 1.0 %
Suitability for determination of aldehydes: passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service