Skip to Content
Merck

104415

SAFC

Oleic acid

EMPROVE® EVOLVE

Synonym(s):

cis-9-Octadecenoic acid, Elainic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1044150010
€601.00
1044150100
€2,888.00

€601.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1044150010
€601.00
1044150100
€2,888.00

About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein:
1726542
MDL number:
UNSPSC Code:
12352211

€601.00


Estimated to ship on25 May 2025


Request a Bulk Order

vapor pressure

1 mmHg ( 176 °C)

product line

EMPROVE® EVOLVE

refractive index

n20/D 1.459 (lit.)

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

application(s)

pharma/biopharma processes

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

ZQPPMHVWECSIRJ-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.
Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service