Skip to Content
Merck
All Photos(1)

Documents

1.04242

Sigma-Aldrich

Fuchsin (C.I. 42510)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN3
CAS Number:
Molecular Weight:
337.85
UNSPSC Code:
41116130
NACRES:
NA.21

form

solid

Quality Level

composition

Dye, ≥80% spectrophotometric assay

pH

5-6 (25 °C, 1 g/L in H2O)

mp

235 °C (decomposition)

solubility

4 g/L

bulk density

500 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C20H19N3.ClH/c1-13-12-16(6-11-19(13)23)20(14-2-7-17(21)8-3-14)15-4-9-18(22)10-5-15;/h2-12,23H,21-22H2,1H3;1H

InChI key

AXDJCCTWPBKUKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This staining dye “Fuchsin (C.I. 42510) - for microscopy Certistain®” is used for staining of acid-fast bacteria (AFB) according to Kinyoun and serves the purpose of the bacteriological and histological investigation of sample material of human origin, for example smears of enriched bacterial cultures or histological sections of e. g. the lung. The cell wall of acid-fast bacteria has a high proportion of wax and lipids and hence absorbs dyes only very slowly. As a measure to enhance the absorption of the fuchsin dye and thus the formation of the mycolate-fuchsin complex in the cell wall, the carbolfuchsin solution applied to the specimen is normally heated to evaporation point. If the Kinyoun carbolfuchsin solution is used, the heating of the solution is dispensable. Consequently, the release of hazardous phenolic vapors will be avoided. Once the acid-fast bacteria have absorbed the fuchsin dye, it is virtually impossible to decolorize them again, even when they are intensively treated with a decolorizing solution such as e. g. hydrochloric acid in ethanol. Accordingly, acid-fast bacteria are termed as acid- and alcohol-fast for staining, and are stained red in the microscopic visualization. Correspondingly, all non-acid-fast microorganisms are counterstained with an appropriate dye. In the present operating instructions is counterstained accordance with methylene blue. Pretreatment of the specimens with Sputofluol dissolves the bacteria from the surrounding viscid sputum and cell material. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

The dye has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining.

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany
SPUTOFLUOL is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service