Skip to Content
Merck
All Photos(1)

Key Documents

1.04119

Osmium(VIII) oxide

for microscopy

Synonym(s):

Osmium(VIII) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O4Os
CAS Number:
Molecular Weight:
254.23
MDL number:
UNSPSC Code:
12352303

vapor pressure

10 hPa ( 20 °C)

Quality Level

potency

15 mg/kg LD50, oral (Rat)

color

yellow

pH

6 (20 °C, 25 g/L in H2O)

bp

130 °C/1013 hPa

mp

40 °C (decomposition)

solubility

65 g/L

density

4.91 g/cm3 at 20 °C

bulk density

1500 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

InChI

1S/4O.Os

InChI key

VUVGYHUDAICLFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Osmium tetroxide for microscopy can be used to visualize unsaturated lipids. A part of the lipids is immediately blackened by the influence of osmic acid solution. The immediate black coloration results from the -C=C- double bond of the unsaturated fatty acid. The fatty acids that are made brownish at first can be blackened by treatment with 60-70% alcohol. For electron microscopy or semithin sectioning the glutardialdehyde-fixed specimens are post-fixed with osmium(VIII) oxide. Lipid inclusions appear black on a yellow to brown background.

Application

for microscopy

Analysis Note

Identity (Raman-spectrum): conforms

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service