Skip to Content
Merck
All Photos(1)

Documents

1.03999

Sigma-Aldrich

Formaldehyde solution min. 37% free from acid

stabilized with about 10% methanol and calcium carbonate for histology

Synonym(s):

Formaline solution, Methanal solution, Methyl aldehyde solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.21

vapor pressure

1.3 hPa

Quality Level

form

liquid

autoignition temp.

300 °C (Formaldehyde)

expl. lim.

7-73 % (v/v) Formaldehyde)

IVD

for in vitro diagnostic use

concentration

37-38% (acidimetric)

color

colorless

pH

2.8-4 (20 °C in H2O)

bp

93-96 °C/1013 hPa

mp

<-15 °C

transition temp

flash point 64 °C

density

1.09 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Formaldehyde solution min. 37% free from acid - stabilized with about 10% methanol and calcium carbonate for histology, is used for human-medical cell diagnosis and serves the purpose of the histological fixation of sample material of human origin. An optimal fixation in the pre-analytical phase of the histological process is one of the most important preconditions for ensuring the good preservation status of the tissue in question and of the structures and macromolecules it contains and thus for enabling a precise histopathological analysis of specimen material. When used properly, this fixing solution is optimally suited for this purpose. The product is a concentrated formaldehyde solution min. 37 %, stabilized, corresponding to a so-called "formalin solution 100 %". The term "100 % formalin solution" is a well-established term used in histological circles for solutions of at least 37 % formaldehyde in water. This has historical reasons. Used in conjunction with other in vitro diagnostic products from our portfolio, in the pre-analytical phase of the histological process it serves the purpose of preserving tissue structures and macromolecules, which are of decisive relevance in histopathological analyses. For more details, please see instructions for use (IFU). This product is registered as IVD and CE marked.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

147.2 °F

Flash Point(C)

64 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service