Skip to Content
Merck
All Photos(1)

Key Documents

1.03887

Supelco

Fluorescein sodium (C.I. 45350)

Fluorescence indicator Reag. Ph Eur

Synonym(s):

Fluorescein sodium (C.I. 45350)

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€53.50
250 G
€238.00

€53.50


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
50 G
€53.50
250 G
€238.00

About This Item

Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
MDL number:
UNSPSC Code:
12171501
EC Index Number:
208-253-0
NACRES:
NB.21

€53.50


Estimated to ship on29 March 2025


Request a Bulk Order

Product Name

Fluorescein sodium (C.I. 45350), indicator Reag. Ph Eur

Agency

reag. Ph. Eur.

Quality Level

form

powder

potency

6720 mg/kg LD50, oral (Rat)

loss

≤10% loss on drying, 110°C

color

red-brown

pH

8.3 (20 °C, 10 g/L in H2O)

mp

>300 °C

solubility

500 g/L

bulk density

600 kg/m3

λmax

490-492 nm (buffer pH 8.0)

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]c1cc2c(cc1)C(=C4C(=CC(=O)C=C4)O2)c3c(cccc3)C(=O)[O-]

InChI

1S/C20H12O5.2Na/c21-11-5-7-15-17(9-11)25-18-10-12(22)6-8-16(18)19(15)13-3-1-2-4-14(13)20(23)24;;/h1-10,21H,(H,23,24);;/q;2*+1/p-2

InChI key

NJDNXYGOVLYJHP-UHFFFAOYSA-L

Analysis Note

Appearance: Redishbrown powder, eventually with granular and bright parts.
Identity (UV/VIS-Spectrum): passes test
Identity (IR-spectrum): passes test
Absorption maximum λmax. (buffer pH 8.0): 490 - 492 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; buffer pH 8.0; calc. on dried substance): ≥ 1850
Loss on drying (110 °C): ≤ 10 %
Suitability as indicator (Adsorption indicator for argentometry): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

423.7 °F - Pensky-Martens closed cup

Flash Point(C)

217.6 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service