Skip to Content
Merck
All Photos(1)

Key Documents

1.02837

Millipore

L-Cystine

for biochemistry

Synonym(s):

L-Cystine, Cys

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€631.00

€631.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€631.00

About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S2
CAS Number:
Molecular Weight:
240.30
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-296-3
NACRES:
NA.21

€631.00


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

form

solid

mp

261-262 °C (decomposition)

solubility

insoluble 0.19 g/L

density

1.677 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

2-30°C

SMILES string

S(SC[C@H]([N+H3])C(=O)[O-])C[C@H]([N+H3])C(=O)[O-]

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

Analysis Note

Assay (bromatometric, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine-crystalline powder
Appearance of solution (0.2 mol/l, hydrochloric acid 1 mol/l): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): -220.0 to - 225.0
Chloride (Cl): ≤ 0.02 %
Sulfate (SO₄): ≤ 0.05 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Zn (Zinc): ≤ 0.0005 %
NH₄ (Ammonium): ≤ 0.01 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.3 %
Loss on drying (105 °C; 3 h): ≤ 0.3 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service