Skip to Content
Merck

Skip To

1.02764

Copper(II) oxide

granular for elementary analysis

Synonym(s):

Copper(II) oxide, Copper monoxide granulated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 g
Check Cart for Availability
€ 405,00

About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
UNSPSC Code:
12352303
EC Index Number:
215-269-1
NACRES:
NA.21
MDL number:

€ 405,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

potency

470 mg/kg LD50, oral (Rat)

pH

7 (20 °C, 50 g/L in H2O, slurry)

mp

1326 °C (decomposition)

density

6.48 g/cm3 at 25 °C

bulk density

500 kg/m3

storage temp.

no temp limit

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Application


  • Facile Synthesis and Electrochemical Characterization of Polyaniline@TiO(2)-CuO Ternary Composite as Electrodes for Supercapacitor Applications.: This article discusses the synthesis and potential use of copper(II) oxide in supercapacitor electrodes, focusing on energy storage enhancements (Boutaleb et al., 2022).

  • Green Fabrication of Nonenzymatic Glucose Sensor Using Multi-Walled Carbon Nanotubes Decorated with Copper (II) Oxide Nanoparticles for Tear Fluid Analysis.: This study introduces a novel approach using copper(II) oxide for the development of glucose sensors, with applications in health monitoring (Asgari Kheirabadi et al., 2022).

  • Physicomechanical and Antimicrobial Characteristics of Cement Composites with Selected Nano-Sized Oxides and Binary Oxide Systems.: Examines the enhancements in cement composites through the incorporation of copper(II) oxide, focusing on strength and antimicrobial properties (Jędrzejczak et al., 2022).

Analysis Note

Assay (complexometric): ≥ 99
Chloride (Cl): ≤ 0.005 %
Total sulfur (as SO₄): ≤ 0.015 %
Total nitrogen (N): ≤ 0.002 %
C (Carbon): ≤ 0.01 %
Bulk density: 240 - 270 g/100 ml
Particle size (0.71-2 mm): ≥ 90 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.02766208841450812
form

solid

form

solid

form

powder

form

powder

mp

1326 °C (decomposition)

mp

1326 °C (decomposition)

mp

-

mp

-

density

6.48 g/cm3 at 25 °C

density

6.48 g/cm3 at 25 °C

density

-

density

-

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

-

storage temp.

-

pH

7 (20 °C, 50 g/L in H2O, slurry)

pH

7 (20 °C, 50 g/L in H2O, slurry)

pH

-

pH

-

Quality Level

100

Quality Level

300

Quality Level

200

Quality Level

100


Still not finding the right product?

Explore all of our products under Copper(II) oxide


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters