Skip to Content
Merck

Skip To

1.02446

Chloroform-D1

deuteration degree min. 99.95% for NMR spectroscopy MagniSolv

Synonym(s):

Chloroform-D1, Deuterochloroform

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 x 0.75 mL
Check Cart for Availability
€ 60,30

About This Item

Empirical Formula (Hill Notation):
CCl3D
CAS Number:
Molecular Weight:
120.38
UNSPSC Code:
12142201
EC Index Number:
212-742-4
NACRES:
NA.22
MDL number:
Isotopic purity:
≥99.95 atom % D
Form:
liquid

€ 60,30


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

211 hPa ( 20 °C)

Quality Level

isotopic purity

≥99.95 atom % D

form

liquid

potency

695 mg/kg LD50, oral (Rat), >3980 mg/kg LD50, skin (Rabbit)

technique(s)

NMR: suitable

impurities

≤0.005% Water (HO) (NMR)

bp

60 °C/1013 hPa

mp

-64.1 °C

density

1.50 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)[2H]

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Application


  • Extraction of alkaloids for NMR-based profiling: exploratory analysis of an archaic Cinchona bark collection.: This study explores the methods of extracting alkaloids for NMR profiling, highlighting the application of Chloroform-D1 in the chemical analysis process to ensure accurate and detailed spectroscopy results. The research was conducted using an archaic Cinchona bark collection, providing insights into the historical and analytical applications of Chloroform-D1 in academic settings (Yilmaz et al., 2012).

Analysis Note

Deuteration degree: ≥ 99.95 %
Water (NMR, HO): ≤ 0.005 %
Performance test (NMR-spectrum): passes test

Other Notes

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.024501.032961.03420
technique(s)

NMR: suitable

technique(s)

NMR: suitable

technique(s)

NMR: suitable

technique(s)

NMR: suitable

isotopic purity

≥99.95 atom % D

isotopic purity

-

isotopic purity

-

isotopic purity

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

liquid

form

liquid

form

liquid

form

liquid

bp

60 °C/1013 hPa

bp

60 °C/1013 hPa

bp

60 °C/1013 hPa

bp

60 °C/1013 hPa

impurities

≤0.005% Water (HO) (NMR)

impurities

≤0.010% Water (H<SUB>2</SUB>O) (NMR)

impurities

≤0.01% Water (H<SUB>2</SUB>O) (NMR), 0.03% Tetramethylsilane (NMR)

impurities

-


Still not finding the right product?

Explore all of our products under Chloroform-D1


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

wgk

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

865-49-6

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
102446002504022536686905
102446000904022536033679
102446001004022536033686
102446010004022536033693

Questions

Reviews

No rating value

Active Filters