Skip to Content
Merck

Skip To

1.02441

Pumice stone

granular

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
11111610


description

for elemental analysis

Quality Segment

form

granular

particle size

0.8-3 mm (≥ 90%)

pH

7.5 -8.0

bulk density

about 50 g/100 mL

storage temp.

no temp limit

SMILES string

CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC

InChI

1S/C28H37FO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3

InChI key

CIWBQSYVNNPZIQ-UHFFFAOYSA-N

General description

granular for elementary analysis

Analysis Note

Substances soluble in dilute hydrochloric acid (as SO₄): ≤ 5.0 %
Loss on ignition (900 °C): ≤ 5.0 %
Bulk density: about 50 g/100ml
Particle size (0.8 - 3 mm): ≥ 90 %


Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters