Skip to Content
Merck
All Photos(1)

Key Documents

1.02384

Supelco

Calcium chloride tetrahydrate

99.995 Suprapur®

Synonym(s):

Calcium chloride tetrahydrate, Calcium dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€273.00

€273.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€273.00

About This Item

Empirical Formula (Hill Notation):
CaCl2 · 4H2O
CAS Number:
Molecular Weight:
183.05
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-140-8
NACRES:
NA.21

€273.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

description

for inorganic trace analysis

Assay

99.995%

form

solid

potency

1000 mg/kg LD50, oral (Rat)
2630 mg/kg LD50, skin (Rat)

pH

5-7 (20 °C, 50 g/L in H2O)

mp

772 °C (anhydrous substance)

storage temp.

2-30°C

SMILES string

[Ca+2].Cl.Cl.[O-H].[O-H].O.O

InChI

1S/Ca.2ClH.4H2O/h;2*1H;4*1H2/q+2;;;;;;/p-2

InChI key

ZNLVAQJGGDVQAU-UHFFFAOYSA-L

Related Categories

Analysis Note

Purity (metallic): ≥ 99.995 %
Assay: ≥ 96 %
Phosphate (PO₄): ≤ 10 ppm
Sulfate (SO₄): ≤ 20 ppm
Al (Aluminium): ≤ 0.01 ppm
Ba (Barium): ≤ 2 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.01 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.01 ppm
Cu (Copper): ≤ 0.005 ppm
Eu (Europium): ≤ 0.01 ppm
Fe (Iron): ≤ 0.05 ppm
K (Potassium): ≤ 2 ppm
La (Lanthanum): ≤ 0.01 ppm
Li (Lithium): ≤ 1 ppm
Mg (Magnesium): ≤ 10 ppm
Mn (Manganese): ≤ 0.01 ppm
Na (Sodium): ≤ 5 ppm
Ni (Nickel): ≤ 0.01 ppm
Pb (Lead): ≤ 0.01 ppm
Rb (Rubidium): ≤ 5 ppm
Sc (Scandium): ≤ 0.01 ppm
Sm (Samarium): ≤ 0.01 ppm
Tl (Thallium): ≤ 0.005 ppm
Y (Yttrium): ≤ 0.01 ppm
Yb (Ytterbium): ≤ 0.01 ppm
Zn (Zinc): ≤ 0.01 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service