Skip to Content
Merck
All Photos(1)

Key Documents

1.02352

Millipore

Cellobiose

for biochemistry

Synonym(s):

Cellobiose

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€224.00

€224.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€224.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
MDL number:
UNSPSC Code:
12352201
EC Index Number:
208-436-5
NACRES:
NA.79

€224.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

pH

5-7 (20 °C, 50 g/L in H2O)

mp

225-230 °C (decomposition)

bulk density

600 kg/m3

storage temp.

15-25°C

SMILES string

O1C(C(C(C(C1CO)O)O)O)OC2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2

InChI key

GUBGYTABKSRVRQ-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 98 %
Identity (IR-spectrum): conforms
Spec. rotation [α²0/D (50 g/l; water): +34.0 - +35.0 °
Heavy metals (as Pb): ≤ 0.001 %
Water (according to Karl Fischer): ≤ 0.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service