Skip to Content
Merck
All Photos(1)

Key Documents

1.02310

SAFC

PARTECK® CCS Croscarmellose Sodium

EMPROVE® ESSENTIAL, Ph. Eur., JP, NF

Pharma Manufacturing

Synonym(s):

PARTECK® CCS Croscarmellose Sodium

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 KG
€133.00
25 KG
€4,510.00

€133.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
0.5 KG
€133.00
25 KG
€4,510.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302

€133.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest a Sample

Agency

JP
NF
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

fine powder

pH

5-7 (20 °C, 10 g/L in H2O)

solubility

30 g/L

application(s)

pharmaceutical
solid formulation

storage temp.

2-30°C

General description

With formulation challenges in mind, we have used particle engineering technologies to develop functional excipients specifically for solid dosage forms. The products in our Parteck® portfolio feature unique particle properties tailored for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® CCS excipient is a superdisintegrant for oral pharmaceutical formulations. Its unique swelling properties shorten tablet disintegration time, thereby increasing disintegration efficiency. Parteck® CCS is highly flexible and suitable for use in capsule formulation, wet granulation, roller compaction and direct compression.

Features and Benefits

  • Fast disintegration
  • Rapid water absorption
  • Good tablet dissolution for a wide variety of applications
  • Does not contain peroxides – for better API stability
  • Flexible in use
  • Very well suited for direct compression, roller compaction, wet granulation and capsule technologies.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service