Skip to Content
Merck
All Photos(1)

Key Documents

1.02274

Supelco

Cerium(IV) sulfate tetrahydrate

for analysis EMSURE®

Synonym(s):

Cerium(IV) sulfate tetrahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€101.00
100 G
€201.00
250 G
€335.00

€101.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€101.00
100 G
€201.00
250 G
€335.00

About This Item

Linear Formula:
Ce(SO4)2 · 4 H2O
CAS Number:
Molecular Weight:
404.30
MDL number:
UNSPSC Code:
12352302
EC Index Number:
237-029-5
NACRES:
NA.21
form:
solid
solubility:
207 g/L

€101.00


Estimated to ship on18 May 2025


Request a Bulk Order

vapor pressure

<1 Pa ( 50 °C)

Quality Level

product line

EMSURE®

Assay

≥98.0% (Ce(SO4)2.4H2O basis, potentiometry)

form

solid

impurities

≤0.005% Substances insoluble in sulfuric acid

pH

0.5 (20 °C, 207 g/L in H2O)

mp

200 °C (Elimination of water of crystallisation)

solubility

207 g/L

density

3.24 g/cm3 at 20 °C

bulk density

650‑850 kg/m3

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.01%

cation traces

Fe: ≤0.005%
heavy metals (as Pb): ≤0.005%

storage temp.

15-25°C

SMILES string

[Ce+3].[S](=O)(=O)([O-])[O-].[S](=O)(=O)([O-])[O-].O.O.O.O.[H+]

InChI

1S/Ce.2H2O4S.4H2O/c;2*1-5(2,3)4;;;;/h;2*(H2,1,2,3,4);4*1H2/q+3;;;;;;/p-3

InChI key

ZBNJQJDKRZKDSV-UHFFFAOYSA-K

Related Categories

Analysis Note

Assay (potentiometric, Ce(SO₄)₂· 4 H₂O): ≥ 98.0 %
Substances insoluble in sulfuric acid: ≤ 0.005 %
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.005 %
Fe (Iron): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepak Kumar Yadav et al.
Nucleic acids research, 48(4), 2091-2106 (2019-12-26)
Staufen1 (STAU1) is a dsRNA binding protein mediating mRNA transport and localization, translational control and STAU1-mediated mRNA decay (SMD). The STAU1 binding site (SBS) within human ADP-ribosylation factor1 (ARF1) 3'UTR binds STAU1 and this downregulates ARF1 cytoplasmic mRNA levels by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service