Skip to Content
Merck

Skip To

1.01809

Volatile Organic Acids Test

photometric, 50-3000 mg/L (acetic acid), Spectroquant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116116
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Spectroquant®

Quality Segment

usage

sufficient for 100 tests

analyte functional class(es)

volatiles 

analyte chemical class(es)

organic acids

measuring range

50-3000 mg/L (acetic acid), 50-3000 mg/L (butyric acid)

technique(s)

photometry: suitable

application(s)

biotechnology/fermenter
disposal drainage water
environmental
wastewater

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100, for use with Spectroquant® Prove 300, for use with Spectroquant® Prove 600

detection method

photometric (Hydroxamic acids / iron(III) salt)

storage temp.

15-25°C

General description

Method: photometric 50 - 3000 mg/l Spectroquant®

This Spectroquant® Volatile Organic Acids Reagent Test allows the accurate determination of the concentration of lower fatty acids (“volatile organic acids”) in aqueous samples.


Method applied: In an acidic medium, lower fatty acids react with a diole to form fatty acid esters, which are subsequently converted into hydroxamic acids with hydroxylamine. These in turn react with iron(III) ions to form red complexes that are determined photometrically.


The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.


All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.017491.192511.14758
detection method

photometric (Hydroxamic acids / iron(III) salt)

detection method

photometric (Hydroxamic acids / iron(III) salt)

detection method

photometric (Iron reduction)

detection method

photometric (Diphenylcarbazide)

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

measuring range

50-3000 mg/L (acetic acid), 50-3000 mg/L (butyric acid)

measuring range

50-3000 mg/L (acetic acid), 50-3000 mg/L (butyric acid)

measuring range

0.020-0.500 mg/L (DEHA), 0.027-0.667 mg/L (Carbohy), 0.05-1.32 mg/L (Hydro), 0.08-1.95 mg/L (ISA), 0.09-2.17 mg/L (MEKO)

measuring range

0.010-3.00 mg/L (Cr), 0.02-6.69 mg/L (CrO4)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Prove 100, for use with Spectroquant® Prove 600, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 300

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

usage

sufficient for 100 tests

usage

sufficient for 25 tests

usage

sufficient for 200 tests

usage

sufficient for 250 tests

analyte chemical class(es)

organic acids

analyte chemical class(es)

organic acids

analyte chemical class(es)

-

analyte chemical class(es)

-


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Kidney

wgk

WGK 2

Storage Class

8A - Combustible corrosive hazardous materials



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters