Skip to Content
Merck
All Photos(1)

Documents

1.01766

Sigma-Aldrich

Benzene-D6

deuteration degree min. 99.95% for NMR spectroscopy MagniSolv

Synonym(s):

Benzene-D6, Hexadeuterobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D6
CAS Number:
Molecular Weight:
84.15
MDL number:
UNSPSC Code:
12142201
EC Index Number:
214-061-8
NACRES:
NA.22

vapor pressure

101 hPa ( 20 °C)

Quality Level

isotopic purity

≥99.95 atom % D

form

liquid

autoignition temp.

555 °C

potency

930 mg/kg LD50, oral (Rat)
>8260 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-8.0 % (v/v)

technique(s)

NMR: suitable

impurities

≤0.005% Water (HO)

kinematic viscosity

0.78 cSt(20 °C)

bp

79 °C/1013 hPa

mp

6.7 °C

transition temp

flash point -11 °C

density

0.95 g/cm3 at 20 °C

storage temp.

no temp limit

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1D,2D,3D,4D,5D,6D

InChI key

UHOVQNZJYSORNB-MZWXYZOWSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Analysis Note

Deuteration degree: ≥ 99.95 %
Water (NMR, HO): ≤ 0.005 %
Performance test (NMR-spectrum): passes test

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service