Skip to Content
Merck
All Photos(1)

Key Documents

1.01729

Supelco

Barium nitrate

for analysis EMSURE® ACS

Synonym(s):

Barium nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€137.00

€137.00


Estimated to ship on07 April 2025


Request a Bulk Order

Select a Size

Change View
500 G
€137.00

About This Item

Linear Formula:
Ba(NO3)2
CAS Number:
Molecular Weight:
261.34
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-020-5
NACRES:
NA.21
Assay:
≥99% (complexometric)
grade:
ACS reagent
form:
solid

€137.00


Estimated to ship on07 April 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

≥99% (complexometric)

form

solid

potency

355 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

5.0-8.0 (25 °C, 50 g/L in H2O)

mp

592 °C

density

3.2 g/cm3 at 20 °C

bulk density

1900 g/m3

anion traces

chloride (Cl-): ≤0.0005%

cation traces

Ca: ≤0.002%
Fe: ≤0.0002%
K: ≤0.005%
Na: ≤0.005%
Sr: ≤0.05%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Ba+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-]

InChI

1S/Ba.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

IWOUKMZUPDVPGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • ′Green firecrackers′ with reduced barium emissions in particulate matter.: This study explores the development of environmentally friendly firecrackers using barium nitrate with reduced particulate matter emissions, contributing to cleaner air. (Wankhede et al., 2023).

  • Hydration Structures on gamma-Alumina Surfaces With and Without Electrolytes Probed by Atomistic Molecular Dynamics Simulations.: The research investigates hydration structures on gamma-alumina surfaces, with barium nitrate serving as one of the electrolytes in the simulations. (Drecun et al., 2022).

  • A novel metal-organic framework of Ba-hemin with enhanced cascade activity for sensitive glucose detection.: This paper presents a novel Ba-hemin metal-organic framework that enhances cascade activity, utilizing barium nitrate in the synthesis process, for sensitive glucose detection. (Yi et al., 2022).

  • Solar light-driven photocatalysis using BaFe2O4/rGO for chlorhexidine digluconate-contaminated water: comparison with artificial UV and visible light-mediated photocatalysis.: The study compares solar light-driven and artificial light-driven photocatalysis for water purification, utilizing barium nitrate in the synthesis of the photocatalyst. (Singh and Mishra, 2022).

  • Structural and dynamic properties of some aqueous salt solutions.: This research examines the structural and dynamic properties of aqueous salt solutions, including barium nitrate, through detailed physical chemistry analysis. (Drecun et al., 2021).

Analysis Note

Assay (complexometric): ≥ 99.0 %
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water; 25 °C): 5.0 - 8.0
Chloride (Cl): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.002 %
Fe (Iron): ≤ 0.0002 %
K (Potassium): ≤ 0.005 %
Na (Sodium): ≤ 0.005 %
Sr (Strontium): ≤ 0.05 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service