Skip to Content
Merck
All Photos(1)

Key Documents

1.01603

Sigma-Aldrich

Gram-Color modified

(phenol-free) staining kit for Gram staining method on bacteriological smears

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€263.00

€263.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
1 SET
€263.00

About This Item

UNSPSC Code:
41116130
NACRES:
NA.74

€263.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

does not contain

phenol

IVD

for in vitro diagnostic use

technique(s)

Gram staining: suitable

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

The Gram-Color modified (phenol-free) - staining kit for Gram staining method on bacteriological smears, is used for human-medical cell diagnosis and serves the purpose of the bacteriological investigation of sample material of human origin from smears of body fluids. Gram-Color modified is a staining kit which is used for a modified Gram staining. The Gram-Color modified solutions are modified and designed in such a way that staining can be carried out on the staining rack. In bacteriology, the Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative. The mureine structure of the bacteria wall is the basis of the color affinity. In the first step, bacteria will be stained with crystal violet an aniline dye. After the treatment with iodine solution (Lugol′s solution), a dye-iodine complex will form. During the decolorizing step, this complex stays in the multilayer mureine structure of the cell wall of Gram-positive bacteria - they will appear blue-violet. Gram-negative bacteria, by contrast, have a cell wall consisting of a single-layered murein structure, and correspondingly re-release the staining dye with the decoloring solution. Gram-negative bacteria will be counterstained with fuchsin solution and will then appear red.
The kit is sufficient for up to 65-70 applications. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy: passes test
gram-positve microorganisms: dark blue
gram-negative microorganisms: red

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F

Flash Point(C)

-10 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service