Skip to Content
Merck
All Photos(1)

Key Documents

1.01506

Supelco

Silver

wool for elemental analysis

Synonym(s):

Silver

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€209.00

€209.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
€209.00

About This Item

Empirical Formula (Hill Notation):
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
EC Index Number:
231-131-3
NACRES:
NA.21

€209.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

bp

2212 °C/1013 hPa

mp

961.9 °C

density

10.5 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Analysis Note

Identity: passes test
Purity: ≥ 99.9 %
Diameter of fibre: about 45 - 55 µm

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service