Skip to Content
Merck
All Photos(1)

Key Documents

1.01091

Supelco

Aluminium hydroxide

powder, hydrargillite EMPLURA®

Synonym(s):

Aluminium hydroxide, Hydrargillite

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€42.50
50 KG
€968.00

€42.50


Available to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€42.50
50 KG
€968.00

About This Item

Linear Formula:
Al(OH)3 · x H2O
CAS Number:
Molecular Weight:
78.00
MDL number:
UNSPSC Code:
12352305
EC Index Number:
244-492-7
NACRES:
NA.21

€42.50


Available to ship on01 April 2025


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

form

powder

loss

30.0-35.0% loss on ignition, 700°C

pH

8-9 (20 °C, 100 g/L in H2O, slurry)

mp

300 °C (Elimination of water of crystallisation)

solubility

water: insoluble at 20 °C

density

2.42 g/cm3 at 20 °C

bulk density

~90 g/100 mL

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.05%

cation traces

Fe: ≤0.01%
Na: ≤0.3%

storage temp.

2-30°C

SMILES string

[Al].O.O.O

InChI

1S/Al.3H2O/h;3*1H2

InChI key

MXRIRQGCELJRSN-UHFFFAOYSA-N

Related Categories

Application


  • Investigation into the Influence of the Process Parameters on the Stability of a Poly(Vinyl)-Alcohol-Based Coating System.: Discusses the stabilization of polymeric materials using aluminium hydroxide, emphasizing its importance in material science for enhancing the durability and performance of coatings (Canale et al., 2024).


Analysis Note

Identity: conforms
Chloride (Cl): ≤ 0.01 %
Sulfate (SO): ≤ 0.05 %
Fe (Iron): ≤ 0.01 %
Na (Sodium): ≤ 0.3 %
Loss on ignition (700°C): 30.0 - 35.0 %
Bulk density: about 90 g/100ml
Particle size (<&#8199;150&#8199;&#181;m): about 90

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service