Skip to Content
Merck
All Photos(2)

Key Documents

1.00991

Supelco

1-Octanol

EMPLURA®

Synonym(s):

1-Octanol, n-Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-917-6
NACRES:
NA.21
bp:
195 °C/1013 hPa
vapor pressure:
0.031 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

0.031 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

270 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.005% Free acid (as C8H16O2)
≤0.1% Water
≤1% Foreign alcohols (GC)

kinematic viscosity

5.6 cSt(40 °C)

bp

195 °C/1013 hPa

transition temp

flash point 86 °C

solubility

0.43 g/L (External MSDS)

density

0.83 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCCCCCCC

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Characterization of Volatile Compounds: This study utilizes 1-Octanol in the analysis of volatile compounds in Valencia orange juice. It highlights its use in the food industry, particularly in quality control and assurance, which is essential for researchers and professionals in the food science sector (Han et al., 2024).

  • Noncontact In Situ Multidiagnostic NMR/Dielectric Spectroscopy: The study discusses the application of 1-Octanol in developing noncontact diagnostic techniques, beneficial for chemists and material scientists involved in analytical and diagnostic methodologies (Morais et al., 2024).

  • Extraction of Bisphenols from Beverages: Highlighting its use in food safety and environmental testing, this study employs 1-Octanol based frameworks for detecting contaminants in consumer products, essential for ensuring public health and safety in the food industry (Cheng et al., 2024).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as C₈H₁₆O₂): ≤ 0.005 %
Density (d 20 °C/ 4 °C): 0.824 - 0.827
Foreign alcohols (GC): ≤ 1.0 %
Readily carbonizable substances: conforms
Water: ≤ 0.1 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service