Skip to Content
Merck
All Photos(3)

Key Documents

1.00845

Supelco

Ethanolamine

for analysis EMSURE®

Synonym(s):

Ethanolamine, 2-Aminoethanol, Monoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€109.00
2.5 L
€178.00

€109.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€109.00
2.5 L
€178.00

About This Item

Empirical Formula (Hill Notation):
C2H7NO
CAS Number:
Molecular Weight:
61.08
MDL number:
UNSPSC Code:
12191502
EC Index Number:
205-483-3
NACRES:
NA.21

€109.00


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

0.5 hPa ( 20 °C)

Quality Level

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

410 °C

potency

1515 mg/kg LD50, oral (Rat)
1025 mg/kg LD50, skin (Rabbit)

expl. lim.

3.4-27 % (v/v)

impurities

≤0.1% Diethanolamine (GC)
≤0.1% Triethanolamine (GC)
≤0.2% Water (Karl Fischer)
≤100 ppb Ba (Barium)
≤20 ppb B (Boron)
≤20 ppb Co (Cobalt)
≤20 ppb Cr (Chromium)
≤20 ppb Cu (Copper)
≤50 ppb Cd (Cadmium)
≤500 ppb Al (Aluminium)
≤500 ppb Ca (Calcium)

pH

12.1 (20 °C, 100 g/L in H2O)

kinematic viscosity

20 cSt(23 °C)

bp

171 °C/1013 hPa

mp

10.5 °C

transition temp

flash point 92.5 °C

density

1.02 g/cm3 at 20 °C

cation traces

Fe: ≤100 ppb
Mg: ≤100 ppb
Mn: ≤20 ppb
Ni: ≤20 ppb
Pb: ≤100 ppb
Sn: ≤100 ppb
Zn: ≤100 ppb

storage temp.

2-30°C

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service