Skip to Content
Merck
All Photos(1)

Key Documents

1.00782

Sigma-Aldrich

Nickel(II) oxide

Technipur®

Synonym(s):

Nickel monoxide, Oxonickel

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€162.00
25 KG
€3,320.00

€162.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€162.00
25 KG
€3,320.00

About This Item

Linear Formula:
NiO
CAS Number:
Molecular Weight:
74.69
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.21

€162.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

density

6.67 g/mL at 25 °C (lit.)

SMILES string

[Ni]=O

InChI

1S/Ni.O

InChI key

GNRSAWUEBMWBQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nickel oxide (NiO) is primarily used in the electronic, optoelectronics, and polymeric industries because of its excellent optical, magnetic, thermal, and mechanical properties. In the semiconductor industry, NiO is used as a p-type intrinsic semiconductor due to its wide bandgap and good chemical stability.[1][2]

Application

Nickel oxide (NiO) is an attractive material because of its many applications in photocatalysis, batteries, capacitor-inductor devices, micro-supercapacitors, electrochromic, and chemical or temperature sensing devices. It can also be used as a conversion reaction-based anode material for lithium-ion batteries due to its low cost, nontoxicity, and high theoretical ability of ~718 mAh/g.[3][2] In perovskite solar cells (PSCs), NiO is commonly used as a hole transport material because of its excellent band alignment and thermal/chemical compatibility with PSCs to enhance the power conversion efficiency of the device.[4]

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance electrospun nanostructured composite fiber anodes for lithium?ion batteries
Chen Y, et al. et al.
Multifunctionality of Polymer Composites , 662-689 (2015)
Nickel Oxide (NiO) Devices and Applications: A Review
Danjumma DS, et al.
International Journal of Engineering Research & Technology (2019)
Influence of Nanostructures in Perovskite Solar Cells
Ghosh P, et al.
Reference Module in Materials Science and Materials Engineering (2016)
Nickel oxide nanoparticles for efficient hole transport in p-i-n and n-i-p perovskite solar cells
Liu, et al.
Journal of Material Chemistry A, 5, 6597-6605 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service