Skip to Content
Merck
All Photos(1)

Key Documents

1.00720

Supelco

Sulfuric acid fuming 65% SO3

Synonym(s):

Sulfuric acid fuming 65% SO3, Oleum

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€809.00

€809.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€809.00

About This Item

Empirical Formula (Hill Notation):
H2O4S · SO3 (1:2)
CAS Number:
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

€809.00


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

105 hPa ( 20 °C)

Quality Level

form

viscous liquid

concentration

64-66% (acidimetric, free SO3)

ign. residue

≤0.02%

color

clear to turbid colorless to brownish

pH

<1 (20 °C, 100 g/L in H2O)

bp

64 °C/1013 hPa

mp

1 °C

density

1.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)O.[S](=O)(=O)=O

InChI

1S/H2O4S.O3S/c1-5(2,3)4;1-4(2)3/h(H2,1,2,3,4);

InChI key

HIFJUMGIHIZEPX-UHFFFAOYSA-N

Analysis Note

Color (visual): colourless to brownish
Appearance of substance (visual): clear to turbid
Content of free SO₃ (acidimetric): 64 - 66 %
Residue on ignition: ≤ 0.02 %
The product may be present as a cloudy viscous liquid and form fine suspended particles

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service