Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
500 mL | Check Cart for Availability | € 300,00 |
About This Item
vapor pressure
59 hPa ( 20 °C)
Quality Segment
form
liquid
expl. lim.
3.5-15 % (v/v) ethanol)
IVD
for in vitro diagnostic use
bp
78.3 °C/1013 hPa (ethanol)
transition temp
flash point 18 °C
density
0.84 g/cm3 at 20 °C
application(s)
clinical testing
diagnostic assay manufacturing
hematology
histology
storage temp.
2-8°C
General description
The resorcin-fuchsin staining method is a regressive method. In tissue sections, nuclei can be stained using a 0.1% nuclear fast red-aluminum sulfate solution (Product number 1.00121) if not only elastic fibers, but also nuclei and the background also need to be visualized.
Brilliant staining results of these fibres are usually achieved when using the ELASTIN staining solution acc. to Weigert - for microscopy in combination with the Picrofuchsin solution acc. van Gieson - for microscopy, Product number 1.00199 and nuclear staining with Weigert′s iron hematoxylin kit - for nuclear staining in histology, Product number 1.15973 respectively Nuclear fast red-aluminium sulfate solution 0.1% - for microscopy, Product number 1.00121.
The 500ml bottle is sufficient for 400 - 500 applications. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Analysis Note
Nuclei: blue-brown
Collagen: red
Muscle fibers: yellow
Cytoplasm: yellow
Elastic fibers: black
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form - | form liquid | form liquid |
| application(s) clinical testing | application(s) clinical testing | application(s) clinical testing | application(s) clinical testing |
| Quality Level 400 | Quality Level 400 | Quality Level 400 | Quality Level 400 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 15-25°C | storage temp. 15-25°C |
| IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use |
| density 0.84 g/cm3 at 20 °C | density - | density 0.80 g/cm3 at 20 °C | density 1.05 g/cm3 at 20 °C |
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
64.4 °F
flash_point_c
18 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





