Skip to Content
Merck
All Photos(1)

Key Documents

1.00480

Supelco

Molybdatophosphoric acid

spray solution for thin-layer chromatography

Synonym(s):

Molybdatophosphoric acid spray solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€92.20

€92.20


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€92.20

About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

€92.20


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

autoignition temp.

425 °C (2-Propanol)

expl. lim.

2-13.4 % (v/v) 2-Propanol()

technique(s)

thin layer chromatography (TLC): suitable

transition temp

flash point 12 °C (2-Propanol)

density

0.84 g/cm3 at 25 °C

storage temp.

15-25°C

General description

Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.

Analysis Note

Application test: passes test

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.6 °F - (2-Propanol)

Flash Point(C)

12 °C - (2-Propanol)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service