Skip to Content
Merck
All Photos(1)

Key Documents

1.00474

Supelco

Phosphate Test, photometric

photometric, 0.05-5.00 mg/L (PO4-P), 0.2-15.3 mg/L (PO43-), 0.11-11.46 mg/L (P2O5), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
€136.00

€136.00


Estimated to ship on23 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
€136.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

€136.00


Estimated to ship on23 May 2025


Request a Bulk OrderRequest more information

Product Name

Phosphate Cell Test, photometric, 0.05-5.00 mg/L (PO4-P), 0.2-15.3 mg/L (PO43-), 0.11-11.46 mg/L (P2O5), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

phosphate

measuring range

0.05-5.00 mg/L (PO4-P)
0.11-11.46 mg/L (P2O5)
0.2-15.3 mg/L (PO43-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Phosphomolybdenum blue)

storage temp.

15-25°C

General description

(o-phosphate) Method: Photometric, 0.05 - 5.00 mg/L (PO4-P), 0.2 - 15.3 mg/L (PO43-), 0.11 - 11.46 mg/L (P2O5), Spectroquant®
The method is analogous to EPA 365.2+3, APHA 4500-P E, and DIN EN ISO 6878.
The Spectroquant®Phosphate Cell Test allows the accurate quantification of phosphate. Elevated levels of phosphate might lead to eutrophication of water bodies, which is a risk to the environment. Therefore, the concentration of phosphate in drinking water and wastewater is regulated in many countries.
Further applications are the determination of phosphate in foods & beverages (e.g. beer) and in environmental and agricultural samples (e.g. soil).
The Spectroquant® Cell Test Kits come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Tests are equipped with the unique Live ID (2D barcode), which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis A Cueto et al.
Environmental science and pollution research international, 27(14), 15984-15993 (2019-05-06)
Phosphorus (P) is a nutrient for plant growth but also a pollutant in water bodies causing eutrophication. The source of P is mainly human and animal wastewater and runoffs from different land uses. The objective of the present study is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service