Skip to Content
Merck

1.00312

SAFC

Hydrochloric acid

25%, EMPROVE® ESSENTIAL, Ph. Helv.

Pharma Manufacturing

Synonym(s):

Hydrochloric acid, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
€51.00
25 L
Inquire
180 L
Inquire

€51.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
€51.00
25 L
Inquire
180 L
Inquire

About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
MDL number:
UNSPSC Code:
12352301

€51.00


Estimated to ship on30 May 2025


Request a Bulk Order

Agency

Ph. Helv.

Quality Level

vapor density

1.3 (vs air)

vapor pressure

12 hPa ( 20 °C)
3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)

product line

EMPROVE® ESSENTIAL

form

liquid

concentration

25%

pH

<1 (20 °C in H2O)

kinematic viscosity

1.73 cSt(20 °C)

bp

>100 °C (lit.)
107 °C/1013 hPa

mp

-52 °C

density

1.12 g/cm3 at 20 °C
1.18 g/L at 20 °C

application(s)

liquid formulation
ophthalmics
pharma/biopharma processes
pharmaceutical
semi-solid formulation

storage temp.

2-25°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Hydrochloric acid can be used as acidifying agent in a variety of pharmaceutical formulations and processes.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service