Skip to Content
Merck

Skip To

1.00202

N,N-Dimethylformamide

for headspace gas chromatography SupraSolv®

Synonym(s):

DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide, DMF, NSC 5356

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.5 L
Check Cart for Availability
€ 156,00
1 L
Check Cart for Availability
€ 217,00
2.5 L
Check Cart for Availability
€ 406,00

About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
UNSPSC Code:
12191502
NACRES:
NA.03
EC Index Number:
200-679-5
MDL number:
Beilstein/REAXYS Number:
605365
Assay:
≥99.8% (GC)
Technique(s):
GC-HS: suitable
Bp:
153 °C (lit.)
Vapor pressure:
2.7 mmHg ( 20 °C)

€ 156,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

according to Ph. Eur., according to USP

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

product line

SupraSolv®

assay

≥99.8% (GC)

form

liquid

autoignition temp.

833 °F

potency

3010 mg/kg LD50, oral (Rat), 1500 mg/kg LD50, skin (Rabbit)

contains

2-Methyl-2-butene as stabilizer

expl. lim.

15.2 %

technique(s)

GC-HS: suitable

impurities

≤0.05% Water

evapn. residue

≤3.0 mg/L

color

APHA: ≤10

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

density

0.944 g/mL (lit.)

storage temp.

2-30°C

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

General description

SupraSolv® high-purity solvents for headspace gas chromatography (HS-GC) are developed particularly for the analysis of residual solvents (RS) in actives, excipients and drug products according to ICH (International Council for Harmonizaton of Technical Requirements for Registration of Pharmaceuticals for Human Use) Guideline Q3C “Impurities: Guideline for Residual Solvents”.

Application

Solvent for many hydrophobic organic compounds.

Features and Benefits

  • high application reliability due to the application test performed on every batch produced
  • application-oriented quality due to specified concentrations of residual solvents as defined by ICH
  • highly pure solvent with extremely low RS concentrations

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.0305333120-M1.03034
Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

200

technique(s)

GC-HS: suitable

technique(s)

-

technique(s)

cell culture | plant: suitable

technique(s)

-

assay

≥99.8% (GC)

assay

≥99.8% (GC)

assay

≥99.8% (GC)

assay

≥99.5% (GC)

form

liquid

form

liquid

form

liquid

form

liquid

expl. lim.

15.2 %

expl. lim.

15.2 %

expl. lim.

15.2 %

expl. lim.

15.2 %

bp

153 °C (lit.)

bp

153 °C (lit.)

bp

152-154 °C, 153 °C (lit.)

bp

153 °C (lit.)


Still not finding the right product?

Explore all of our products under N,N-Dimethylformamide


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

135.5 °F - closed cup

flash_point_c

57.5 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

68-12-2

CAS No.

68-12-2

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters