Skip to Content
Merck
All Photos(2)

Key Documents

09-893

Sigma-Aldrich

Anti-BAF (BANF1) Antibody

serum, from rabbit

Synonym(s):

53 kDa BRG1-associated factor A, Actin-related protein Baf53a, BAF complex 53 kDa subunit, BRG1-associated factor, INO80 complex subunit K, actin-like 6A, actin-related protein 4, hArpN beta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€497.00

€497.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€497.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€497.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BANF1(8815)

General description

Barrier-to-auto integration factor (BAF or BANF1) plays fundamental roles in nuclear assembly, chromatin organization, gene expression and development. BAF may potently compress chromatin structure and be involved in membrane recruitment and chromatin decondensation during nuclear assembly. BAF contains 2 non-specific dsDNA-binding sites which may promote DNA cross-bridging. BAF is exploited by retroviruses for inhibiting self-destructing autointegration of retroviral DNA, thereby promoting integration of viral DNA into the host chromosome. BAF is found in both the nucleus and cytoplasm and is significantly enriched at the nuclear inner membrane, diffused throughout the nucleus during interphase and concentrated at the chromosomes during the M-phase.

Specificity

This antibody recognizes BAF (BANF1).

Immunogen

KLH-conjugated linear peptide corresponding to human BAF (BANF1).

Application

Use Anti-BAF (BANF1) Antibody (Rabbit Polyclonal Antibody) validated in WB to detect BAF (BANF1) also known as 53 kDa BRG1-associated factor A.
Western Blot Analysis: A 1:1,000 dilution from a previous lot detected BAF in C2C12, Hek293, HeLa, HepG2, Huvec, Jurkat, L6, NIH/3T3, PC12, PC3, and RAW264.7 cell lysates.

Quality

Evaluated by Western Blot in HeLa cell lysate.
Western Blot Analysis: A 1:1,000 dilution of this antibody detected BAF in 10 µg of HeLa cell lysate.

Target description

~10 kDa

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Cheng Yang et al.
F1000Research, 3, 115-115 (2014-09-02)
NKX3.1 is a homeobox transcription factor whose function as a prostate tumor suppressor remains insufficiently understood because neither the transcriptional program governed by NKX3.1, nor its interacting proteins have been fully revealed. Using affinity purification and mass spectrometry, we have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service