Skip to Content
Merck

Skip To

09-874

Anti-SAP30 (yeast) Antibody

from rabbit

Synonym(s):

Sin3 corepressor complex subunit SAP30, Sin3-associated polypeptide, 30 kDa, Sin3-associated polypeptide, 30kDa, Sin3A-associated protein, 30kDa, histone deacetylase complex subunit SAP30

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 431,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
6

€ 431,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

Saccharomyces cerevisiae

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Saccharomyces cerevisiae ... Sap30(855305)

General description

SAP30 (Sin3A-associated protein) is a widely expressed nuclear protein that is functionally conserved between human and yeast. It is a component of the multiprotein histone deacetylase complex containing the transcriptional repressor Sin3, multiple histone deacetylases (HDAC1 and HDAC2), histone binding proteins (RbAp46 and RbAp48) and other polypeptides. SAP30 can recruit the deacetylase complex to promoter DNA in a Sin3 independent manner and is involved in the recruitment of the mSin3 complex to a specific subset of N-CoR corepressor complexes. The latter function suggests a role for SAP30 in N-CoR-mediated repression. Furthermore, SAP30 may be involved in imparting target specificity to the deacetylase complex.
~27 kDa

Immunogen

Epitope: a.a. 141-157
Immunogen was a synthetic peptide corresponding to a.a. 141-157 of yeast SAP30, conjugated to KLH.

Application

Anti-SAP30 (yeast) Antibody is a Rabbit Polyclonal Antibody for detection of SAP30 (yeast) also known as Sin3 corepressor complex subunit SAP30 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Chromatin Biology

Biochem/physiol Actions

Not tested in other species.
Recognizes yeast SAP30 at ~27 kDa.

Physical form

Format: Purified
Protein A purified IgG in 0.1M Tris-Glycine (pH7.4) 150 mM NaCl with 0.05% NaN3.

Preparation Note

Stable for 1 year at 2-8°C from date of shipment.

Analysis Note

Control
Yeast extract
Western Blot Analysis:
1 μg/ml of this antibody detected SAP30 in a yeast extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
04-1549WH0008819M206-913-I
clone

polyclonal

clone

1YO 1B10, monoclonal

clone

4D2, monoclonal

clone

polyclonal

biological source

rabbit

biological source

mouse

biological source

mouse

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

Saccharomyces cerevisiae

species reactivity

mouse, rat, human

species reactivity

human

species reactivity

human, mouse

antibody form

purified immunoglobulin

antibody form

ascites fluid

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

UniProt accession no.

P38429

UniProt accession no.

Q921M3

UniProt accession no.

O75446

UniProt accession no.

Q60520


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters