Skip to Content
Merck
All Photos(1)

Documents

09-818

Sigma-Aldrich

Anti-CARM1 Antibody

from rabbit

Synonym(s):

Protein arginine N-methyltransferase 4, coactivator-associated arginine methyltransferase 1, coactivator-associated arginine methyltransferase-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

species reactivity (predicted by homology)

mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CARM1(10498)
mouse ... Carm1(59035)

General description

CARM1 (coactivator-associated arginine methyltransferase 1) is an arginine N-methyltransferase that catalyzes the transfer of a methyl group from S-adenosyl-L-methionine to arginine residues within proteins. CARM1 targets include methylation of histone H3 at Arg17 and p300 at Arg2142. CARM1 is recruited to promoters upon gene activation together with histone acetyltransferases from the p300 and p160 families. During monocyte inflammatory stimulation, CARM1 acts together with p300 as a coactivator for NF-kB. It is responsible for moving cells toward the inner cell mass in developing blastocysts. CARM1 also plays an important role in androgen receptors and may play a role in prostate cancer progression.

Specificity

Recognizes CARM1, Mr ~60 kDa.

Immunogen

Epitope: a.a. 595-608
KLH-conjugated, synthetic peptide containing the sequence (CSPMSIPTNTMHYGS), corresponding to a.a. 595-608 of human CARM1.

Application

Anti-CARM1 Antibody is a rabbit polyclonal antibody for detection of CARM1 also known as Protein arginine N-methyltransferase 4, coactivator-associated arginine methyltransferase 1 & has been validated in WB.
Western Blot Analysis:
A 1 μg/mL dilution of this lot detected CARM1 in HeLa nuclear extracts. (Figure A).

Target description

~60 kDa

Physical form

Format: Purified
Purified by peptide affinity chromatography. Supplied at 1 mg/ml in 0.1 M Tris-Glycine (pH7.4) 15 mM NaCl, 0.05% sodium azide.

Analysis Note

Control
Included Positive Antigen Control: Catalog # 12-309, HeLa nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria-Elena Torres-Padilla et al.
Nature, 445(7124), 214-218 (2007-01-12)
It has been generally accepted that the mammalian embryo starts its development with all cells identical, and only when inside and outside cells form do differences between cells first emerge. However, recent findings show that cells in the mouse embryo
Heng Hong et al.
Cancer, 101(1), 83-89 (2004-06-29)
Coactivator-associated arginine methyltransferase 1 (CARM1) is a transcriptional coactivator of the androgen receptor (AR). It is involved in the regulation of the biologic functions of the AR. It remains to be determined whether CARM1 is involved in prostatic carcinogenesis. The
D Chen et al.
Science (New York, N.Y.), 284(5423), 2174-2177 (1999-06-26)
The p160 family of coactivators, SRC-1, GRIP1/TIF2, and p/CIP, mediate transcriptional activation by nuclear hormone receptors. Coactivator-associated arginine methyltransferase 1 (CARM1), a previously unidentified protein that binds to the carboxyl-terminal region of p160 coactivators, enhanced transcriptional activation by nuclear receptors
Leila J Mady et al.
Cells, 12(10) (2023-07-06)
1,25-Dihydroxyvitamin D3 (1,25(OH)2D3), the hormonally active form of vitamin D, activates the nuclear vitamin D receptor (VDR) to mediate the transcription of target genes involved in calcium homeostasis as well as in non-classical 1,25(OH)2D3 actions. In this study, CARM1, an
Adam Burton et al.
Cell reports, 5(3), 687-701 (2013-11-05)
Cell plasticity or potency is necessary for the formation of multiple cell types. The mechanisms underlying this plasticity are largely unknown. Preimplantation mouse embryos undergo drastic changes in cellular potency, starting with the totipotent zygote through to the formation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service