Skip to Content
Merck
All Photos(2)

Key Documents

09-718

Sigma-Aldrich

Anti-TGIF2 Antibody

Upstate®, from rabbit

Synonym(s):

TGFB-induced factor homeobox 2, transcription growth factor-beta-induced factor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€316.00

€316.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€316.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€316.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes TGIF2

Immunogen

Epitope: a.a. 189-203
KLH conjugated synthetic peptide (C)-EQDKEDFSSFQLLVE corresponding to a.a. 189-203 of human TGIF.

Application

Anti-TGIF2 Antibody is a Rabbit Polyclonal Antibody for detection of TGIF2 also known as transcription growth factor-beta-induced factor 2 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Routinely evaluated by immunoblot.

Target description

28 kDa

Physical form

ImmunoAffinity Purified
Immunoaffinity purified rabbit IgG in PBS, 0.1% sodium azide, 30% glycerol

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob L Studt et al.
MethodsX, 7, 100791-100791 (2020-02-06)
Measuring total nitrogen, nitrate, and nitrite is critical for compliance with water safety standards. Previous methods for measuring total nitrogen were hazardous, time consuming, and expensive. Here we report a method for measuring total nitrogen in water and soil using
Jing Y Krzeszinski et al.
Nature, 512(7515), 431-435 (2014-07-22)
Bone-resorbing osteoclasts significantly contribute to osteoporosis and bone metastases of cancer. MicroRNAs play important roles in physiology and disease, and present tremendous therapeutic potential. Nonetheless, how microRNAs regulate skeletal biology is underexplored. Here we identify miR-34a as a novel and
F Morgan-Sagastume et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 69(1), 177-184 (2014-01-18)
Polyhydroxyalkanoates (PHAs) are biodegradable polyesters with comparable properties to some petroleum-based polyolefins. PHA production can be achieved in open, mixed microbial cultures and thereby coupled to wastewater and solid residual treatment. In this context, waste organic matter is utilised as
Arka Banerjee et al.
Nucleic acids research, 52(15), 9028-9048 (2024-07-23)
Increasingly many studies reveal how ribosome composition can be tuned to optimally translate the transcriptome of individual cell types. In this study, we investigated the expression pattern, structure within the ribosome and effect on protein synthesis of the ribosomal protein
Brice Korkmaz et al.
Nature protocols, 3(6), 991-1000 (2008-06-10)
The neutrophil serine proteases (NSPs) elastase, proteinase 3 and cathepsin G are multifunctional proteases involved in pathogen destruction and the modulation of inflammatory processes. A fraction of secreted NSPs remains bound to the external plasma membrane, where they remain enzymatically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service