Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IHC, IP, WB
Citations:
1
Pricing and availability is not currently available.
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, rat
technique(s)
ELISA: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PIK3CA(5290)
rat ... Pik3Ca(170911)
General description
110 kDa
Phosphoinositol-3-kinases (PI3 Kinase, PI3K) are a family of lipid kinases that mediate many intracellular signaling responses in both physiological and pathophysiological states. PI3 Kinase is a heteroduplex with three classes (I, II, & III) of catalytic domain. The class I PI3 Kinases are made up of a p85 regulatory protein and a p110 catalytic domain. There are 4 isoforms of the class I p110 catalytic domains. Class I(A) include α, β, γ and class I(B). They are activated downstream via receptor tyrosine kinases or G-protein coupled receptors. Once activated, PI3K generates phosphatidylinositol 3,4,5-trisphosphate (PIP3) from PIP2. It was later found that PTEN, a lipid phosphatase, acts opposite to PI3K by dephosphorylating PIP3, converting it to PIP2, thus de-activating the effects of PI3K. Increased levels of PIP3 in the cell leads to the activation of many key signaling regulators including Akt and PDK as well as calcium release. One of the primary roles of PI3K is its regulation of cell growth and cell metabolism. Both of these play a critical role in cancer as some believe that a tumor’s shift to glycolysis is due to the activation of PI3K. PI3 kinases have been linked with numerous disease states, including cancer, diabetes, allergic response, hypertension, cardiac contractility, atherosclerosis, sepsis and autoimmune/ inflammatory disorders.
Immunogen
Epitope: Internal
KLH-conjugated synthetic linear peptide corresponding to an internal region of class I catalytic domain of PI3 Kinase that is specific to the p110 alpha isoform.
Application
Detect PI3 Kinase using this Anti-PI3 Kinase Antibody, p110α validated for use in WB, IP, IH, ELISA.
Research Category
Signaling
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling
PI3K, Akt, & mTOR Signaling
Biochem/physiol Actions
Recognizes PI3 Kinase p110α.
Physical form
Immunogen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl, and 0.05% sodium azide.
Preparation Note
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl, and 0.05% sodium azide.
Stable for 1 year at 2-8ºC from date of receipt.
Analysis Note
Evaluated by western blot on Jurkat cell extract.
Western Blotting Analysis: 0.5 µg/mL (1:2,000) dilution of this antibody detected PI3K, p110α in Jurkat cell extract.
Western Blotting Analysis: 0.5 µg/mL (1:2,000) dilution of this antibody detected PI3K, p110α in Jurkat cell extract.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-399
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.