Skip to Content
Merck
All Photos(1)

Documents

07-909

Sigma-Aldrich

Anti-phospho-Src (Tyr418) Antibody

Upstate®, from rabbit

Synonym(s):

c-Src, p60-Src Src kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

chicken, human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr418)

Gene Information

human ... SRC(6714)

Specificity

Predicted to cross-react with Rat and Frog (100% homologous) based on sequence homology.
Recognizes Src phosphorylated on Tyrosine residue 418.

Immunogen

Peptide corresponding to amino acid region encompassing the human, mouse, rat, chicken, and frog phospho-Src (Tyr418)

Application

Anti-phospho-Src (Tyr418) Antibody detects level of phospho-Src (Tyr418) & has been published & validated for use in IC, IH & WB.
Immunoblot Analysis: 1:1000 dilutions of this lot detected phospho-Src (Tyr418) in RIPA lysate of Src transfected CEF cells.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by immunoblot.

Target description

60 kDa

Physical form

Affinity Purified rabbit polyclonal IgG in PBS (without Mg2+ and Ca2+), pH 7.3 containing 0.5 mg/mL BSA (IgG, protease free) and 0.025% sodium azide and 50% glycerol.
ImmunoAffinity Purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Chick embryo fibroblast (CEF) cells expressing human wild-type Src protein

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

c-Src and cooperating partners in human cancer
Ishizawar, Rumey and Parsons, Sarah J
Cancer Cell, 6, 209-214 (2004)
Shinkyu Choi et al.
Antioxidants & redox signaling, 30(4), 505-519 (2018-01-18)
Altered redox state has been related to the development of normal pregnancy (NP) and preeclampsia (PE). Endothelial KCa2.3 and KCa3.1 (KCas) play an important role in vasodilation, and KCas levels are affected by oxidative stress. We investigated the mechanisms of
Aida Habibovic et al.
JCI insight, 1(18), e88811-e88811 (2016-11-05)
Chronic inflammation with mucous metaplasia and airway remodeling are hallmarks of allergic asthma, and these outcomes have been associated with enhanced expression and activation of EGFR signaling. Here, we demonstrate enhanced expression of EGFR ligands such as amphiregulin as well
Functional stability of a mixed microbial consortium producing PHA from waste carbon sources.
Erik R Coats,Frank J Loge,William A Smith,David N Thompson,Michael P Wolcott
Applied Biochemistry and Biotechnology null
Src: regulation, role in human carcinogenesis and pharmacological inhibitors.
Tsygankov, A Y and Shore, S K
Current Pharmaceutical Design, 10, 1745-1756 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service